Z740469
MICROGARD™ 2000 coveralls for protection from infective agent
size M (elasticated ankles)
All Photos(1)
About This Item
UNSPSC Code:
46181503
NACRES:
NB.35
Recommended Products
size
M (elasticated ankles)
manufacturer/tradename
Arco 1762004
General description
MICROGARD™ 2000 fully elasticated coverall with hood , setting the standard in Type 5/6 performance and protection, offers protection from infective agents, pesticides, particulates, fibres and liquid chemical spray.
Features:
Approved to:
EN ISO 13982-1 Type 5 Particulate protection
EN13034 Type 6 Reduced spray
EN1073-2 Barrier to radioactive particulates - Class 2
EN14126 Barrier to infective agents
EN1149-5 Anti-static
Features:
- 3-piece hood
- 2-way front zipper with re-sealable storm flap
- Finger loops
- Elasticated hood, waist, wrists and ankles
Approved to:
EN ISO 13982-1 Type 5 Particulate protection
EN13034 Type 6 Reduced spray
EN1073-2 Barrier to radioactive particulates - Class 2
EN14126 Barrier to infective agents
EN1149-5 Anti-static
Legal Information
Microgard is a trademark of Microgard Limited