PZ0406
PF-06815345
≥98% (HPLC)
Synonym(s):
(1S)-1-(5-(4-(4-((3-Chloropyridin-2-yl)((R)-piperidin-3-yl)carbamoyl)-2-fluorophenyl)-1-methyl-1H-pyrazol-5-yl)-1H-tetrazol-1-yl)ethyl ethyl carbonate hydrochloride, Ethyl (S)-1-{5-[4-(4-{(3-chloropyridin-2-yl)[(3R)-piperidin-3-yl]carbamoyl}-2-fluorophenyl)-1-methyl-1H-pyrazol-5-yl]-1H-tetrazol-1-yl}ethyl carbonate hydrochloride, PF 06815345, PF06815345
About This Item
Recommended Products
Quality Level
Assay
≥98% (HPLC)
form
powder
optical activity
[α]/D -34 to -43°, c = 0.3 in methanol
color
white to beige
solubility
DMSO: 2 mg/mL, clear
storage temp.
room temp
SMILES string
Fc1c(ccc(c1)C(=O)N([C@H]5CNCCC5)c4ncccc4Cl)c2c([n](nc2)C)c3[n](nnn3)[C@@H](OC(=O)OCC)C.Cl
InChI key
RFEUFHPTEFEGDQ-KUGOCAJQSA-N
Biochem/physiol Actions
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service