Skip to Content
Merck
All Photos(11)

Key Documents

HPA001525

Sigma-Aldrich

Anti-AHSG antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-α-2-HS-glycoprotein precursor antibody produced in rabbit, Anti-α-2-Z-globulin antibody produced in rabbit, Anti-Ba-α-2-Glycoprotein antibody produced in rabbit, Anti-Fetuin-A antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

RNAi knockdown
independent
independent
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

GLIYRQPNCDDPETEEAALVAIDYINQNLPWGYKHTLNQIDEVKVWPQQPSGELFEIEIDTLETTCHVLDPTPVARCSVRQLKEHAVEGDCDFQLLKLDGKFSVVYAKCDSSPDSAEDVRKVCQDCPLLAPLNDTRVVHAAKAALA

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... AHSG(197)

Looking for similar products? Visit Product Comparison Guide

Immunogen

α-2-HS-glycoprotein precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

AHSG (α-2-HS-glycoprotein) gene encodes a glycoprotein that is present in the serum. It is synthesized by hepatocytes. The gene is localized to human chromosome 3q27, a susceptibility locus for type 2 diabetes and the metabolic syndrome. It may be involved in the regulation of postprandial glucose disposal, insulin sensitivity, weight gain, and fat accumulation. It may serve as a potential therapeutic target in the treatment of type 2 diabetes, obesity, and other insulin-resistant conditions. It is an antagonist of transforming growth factor β, regulating cytokine-dependent osteogenesis. It inhibits insulin receptor tyrosine kinase and some protease activities.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST84501

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Markus Ketteler et al.
Lancet (London, England), 361(9360), 827-833 (2003-03-19)
Vascular calcification is the most prominent underlying pathological finding in patients with uraemia, and is a predictor of mortality in this population. Fetuin-A (alpha2-Heremans Schmid glycoprotein; AHSG) is an important circulating inhibitor of calcification in vivo, and is downregulated during
Suresh T Mathews et al.
Diabetes, 51(8), 2450-2458 (2002-07-30)
Fetuin inhibits insulin-induced insulin receptor (IR) autophosphorylation and tyrosine kinase activity in vitro, in intact cells, and in vivo. The fetuin gene (AHSG) is located on human chromosome 3q27, recently identified as a susceptibility locus for type 2 diabetes and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service