CLS357535
Corning® Falcon® Serological Pipettes
capacity 25 mL, individually wrapped, extended, clear plastic, sterile
Synonym(s):
pipet
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Recommended Products
material
polystyrene
sterility
sterile
feature
individually wrapped extended, clear plastic
subdivision
manufacturer/tradename
Corning 357535
capacity
25 mL
Looking for similar products? Visit Product Comparison Guide
Application
Liquid Handling
Legal Information
Corning is a registered trademark of Corning, Inc.
Falcon is a registered trademark of Corning, Inc.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service