Skip to Content
Merck
All Photos(1)

Key Documents

GP2d

NOTE: Both the cell line and DNA from the cell line may beavailable for this product. Please choose -1VL or VIAL for cells, or -DNA-5UGfor DNA, 95090714, human colon, Epithelial

Synonym(s):

GP2d Cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

biological source

human colon

packaging

tube of 5 μg 95090714-DNA-5UG
pkg of vial of cells 95090714-1VL

growth mode

Adherent

karyotype

Diploid

morphology

Epithelial

products

Not specified

receptors

Epidermal growth factor (EGF)

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human Caucasian colon adenocarcinoma

Cell Line Description

GP2d has been established from the same adenocarcinoma as GP5d (Sigma Catalogue number. 95090715). This has been confirmed by STR profiling at ECACC. The cells were derived from a local recurrence of Duke′s grade B, poorly differentiated carcinoma of the colon from a 71 year old female at surgical resection. The patient received no preoperative chemo- or radiotherapy and had a strong family history of colon cancer with two first degree relatives dying of the disease below the age of 55 years. Both clones possess genetic changes consistent with the pattern of tumour progression in colon cancer but are morphologically distinct. An interstitial deletion on chromosome 5 (region 14q to 22q) and an inverted duplication of bands 10q11 and 10q21 has been reported. Both cell lines contain a normal copy of the ki-ras gene and a copy with a transition in codon 12. GP2d cells show growth response to EGF, TGF α or insulin with an increase in cell numbers. Amphiregulin mRNA is abundant in GP2d but almost undetectable in GP5d.

Application

GP2d has been used to extract genomic DNA for digital PCR (dPCR). It has also been used to extract DNA from the cell with PIK3CA mutation c.3140A>T.

Packaging

NOTE: Both the cell line and DNA from the cell line may beavailable for this product. Please choose -1VL or VIAL for cells, or -DNA-5UGfor DNA.

DNA Profile

STR-PCR Data: Amelogenin: X
CSF1PO: 9,12
D13S317: 8,12
D16S539: 11,13
D5S818: 11,12
D7S820: 9,11
THO1: 7,9.3
TPOX: 8,11
vWA: 17,18

Culture Medium

DMEM + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:6 i.e. seeding at 5x10,000 to 1x100,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service