AXYMCT200NC
Microtubes
no cap, size 2.0 mL, clear, pkg of 10x500tubes/cs
Synonym(s):
centrifuge tubes
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
conical bottom tube
polypropylene
sterility
non-sterile
feature
RNase and DNase free
endotoxin free
graduations
manufacturer/tradename
Axygen Inc MCT-200-NC
Corning
packaging
pkg of 10x500tubes/cs
size
2.0 mL
color
clear
clear
suitability
suitable for (centrifugation)
Looking for similar products? Visit Product Comparison Guide
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service