42981
Legionella Growth Supplement (Twin Pack)
suitable for microbiology
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41171614
NACRES:
NA.85
Recommended Products
sterility
sterile
form
liquid
shelf life
limited shelf life, expiry date on the label
application(s)
environmental
microbiology
storage temp.
2-8°C
suitability
Legionella spp.
Related Categories
Application
A chemical enrichment supplement recommended for enhancing growth of Legionella species.
Packaging
one package contains 5 vials part A and 5 vials part B.
Components
(per vial, sufficient for 500 ml medium)
Legionella Growth Supplement Part A (Cat. No.63958):
L-Cysteine hydrochloride 200.0 mg
Legionella Growth Supplement Part B (Cat. No. 93164) :
Ferric pyrophosphate, soluble 125.0 mg
Sodium selenite 5.0 mg
Distilled water 5.0 ml
Legionella Growth Supplement Part A (Cat. No.63958):
L-Cysteine hydrochloride 200.0 mg
Legionella Growth Supplement Part B (Cat. No. 93164) :
Ferric pyrophosphate, soluble 125.0 mg
Sodium selenite 5.0 mg
Distilled water 5.0 ml
Storage Class Code
10 - Combustible liquids
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service