Skip to Content
Merck
All Photos(1)

Key Documents

30374

Millipore

UTI ID Membrane

suitable for microbiology, white colored, sterile, diameter 70 mm

Synonym(s):

Indicator membrane, Urinary Tract Infections ID Membrane

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171621
NACRES:
NA.85

membrane size

70 mm

sterility

sterile

shelf life

limited shelf life, expiry date on the label

storage condition

protect from light

technique(s)

microbe id | specific enzyme detection: suitable

application(s)

clinical testing
food and beverages

microbiology

storage temp.

2-8°C

suitability

Enterococcus spp.
Escherichia coli
Klebsiella spp.
Proteus spp.
Pseudomonas spp.
Staphylococcus spp.
coliforms

General description

UTI ID Membranes are substrate-carrying membranes used for rapid detection and confirmation of microorganisms mainly causing urinary tract infections in clinical samples through characteristic color reactions by placing on the agar surface post-incubation. The membranes contain chromogenic substrates and indicators, which serve as the basis for microbe differentiation by color. The target organisms possess enzyme systems that metabolize the substrates in the membrane, leading to a change of color. The colors can be visually detected on the membrane. This indicates microbial enzymatic activity, which serves to identify the genus and species.

Application

UTI ID Membrane is recommended for rapid detection and confirmation of microorganisms mainly causing urinary tract infections. For e.g., E. coli, Proteus, Klebsiella, Pseudomonas, Staphylococcus aureus, and Enterococcus species. They find their application in various sectors, such as pharmaceutical laboratory testing and clinical diagnostics.

Analysis Note

Escherichia coli (25922)pink-purpleStaphylococcus aureus (25923)golden yellowPseudomonas aeruginosa (27853)colorlessEnterococcus faecalis (29212)blue – blue green (small)Klebsiella pneumoniae (13883)blue to purple mucoidProteus mirabilis (12453)Light brown

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service