Skip to Content
Merck
All Photos(2)

Key Documents

ABE993

Sigma-Aldrich

Anti-SLUG (SNAI2)

from rabbit

Synonym(s):

Zinc finger protein SNAI2, Neural crest transcription factor Slug, Protein snail homolog 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

species reactivity (predicted by homology)

rat (based on 100% sequence homology), canine (based on 100% sequence homology), sheep (based on 100% sequence homology), bovine (based on 100% sequence homology), porcine (based on 100% sequence homology)

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... SNAI2(6591)

General description

Zinc finger protein SNAI2 (UniProt: O43623; also known as Neural crest transcription factor Slug, SLUG, Protein snail homolog 2) is encoded by the SNAI2 (also known as SLUG, SLUGH) gene (Gene ID: 6591) in human. SLUG is a transcriptional repressor that modulates both activator-dependent and basal transcription. It is expressed in most adult human tissues, but is not detected in peripheral blood leukocyte. It is observed in discrete foci in interphase nuclei and these nuclear foci do not overlap with the nucleoli, the SP100 and the HP1 heterochromatin or the coiled body, suggesting its association with active transcription or active splicing regions. SLUG is shown to repress BRCA2 expression by binding to its E2-box-containing silencer and recruiting CTBP1 and HDAC1 in breast cells. It contains a SNAG domain (aa 1-20) that resembles a histone H3-like structure and functions as a molecular hook for recruiting LSD1 to repress gene expression in metastasis. SLUG is shown to be involved in the generation and migration of neural crest cells. It also plays a role in mediating RAF1-induced transcriptional repression of the TJ protein, occludin (OCLN) and subsequent oncogenic transformation of epithelial cells. In epidermal keratinocytes, it binds to the E-box in ITGA3 promoter and represses its transcription. SLUG is also involved in the regulation of ITGB1 and ITGB4 expression and cell adhesion and proliferation in epidermal keratinocytes. It can bind to E-box2 domain of BSG and activate its expression during TGFB1-induced epithelial-mesenchymal transition (EMT) in hepatocytes. SLUG phosphorylation by GSK-3 beta results in cytoplasmic localization and degradation. Mutation in SNAI2 gene can cause Waardenburg syndrome 2D that is characterized by sensorineural deafness, pigmentary disturbances, and absence of dystopia canthorum. (Ref.: Lin, Y et al. (2010). EMBO J. 29(11); 1803-16).

Specificity

This rabbit polyclonal antibody detects SLUG/SNAI2 in human cells. It targets an epitope within 12 amino acids from the N-terminal half.

Immunogen

Epitope: unknown
KLH-conjugated linear peptide corresponding to 12 amino acids from the N-terminal half of human Zinc finger protein SNAI2.

Application

Anti-SLUG (SNAI2), Cat. No. ABE993, is a highly specific rabbit polyclonal antibody that targets Zinc finger protein SNAI2 and has been tested for use in Immunohistochemistry (Paraffin) and Western Blotting.
Immunohistochemistry Analysis: A 1:250 dilution from a representative lot detected SLUG (SNAI2) in human bone marrow and human pancreas tissues.
Research Category
Epigenetics & Nuclear Function

Quality

Evaluated by Western Blotting in HepG2 cell lysate.

Western Blotting Analysis: 2 µg/mL of this antibody detected SLUG (SNAI2) in 10 µg of HepG2 cell lysate.

Target description

~31 kDa observed; 29.99 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Affinity Purified
Format: Purified
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dacheng Ding et al.
The Tohoku journal of experimental medicine, 263(3), 199-210 (2024-05-27)
Glioblastoma (GBM) is a malignant primary brain tumor and an essential contributor to morbidity and mortality globally. Arsenic trioxide (ATO) exerts specific roles in preventing tumor growth. This study investigated the role of ATO in GBM cell behaviors and stemness.
Hemin-Induced Endothelial Dysfunction and Endothelial to Mesenchymal Transition in the Pathogenesis of Pulmonary Hypertension Due to Chronic Hemolysis.
Gonzales, et al.
International Journal of Molecular Sciences, 23 (2023)
Kai-Yen Peng et al.
Frontiers in oncology, 11, 716055-716055 (2021-09-28)
Over 90% of colorectal cancer (CRC) patients have mutations in the Wnt/β-catenin pathway, making the development of biomarkers difficult based on this critical oncogenic pathway. Recent studies demonstrate that CRC tumor niche-stromal cells can activate β-catenin in cancer-initiating cells (CICs)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service