Skip to Content
Merck
All Photos(1)

Key Documents

8.20533

Sigma-Aldrich

2,3-Diphenyl-2-cyclopropen-1-one

for synthesis

Synonym(s):

2,3-Diphenyl-2-cyclopropen-1-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H10O
CAS Number:
Molecular Weight:
206.24
MDL number:
UNSPSC Code:
12352115
EC Index Number:
212-948-4
NACRES:
NA.22
form:
powder
Assay:
≥98.0% (HPLC)

Quality Level

Assay

≥98.0% (HPLC)

form

powder

mp

118-122 °C

storage temp.

2-30°C

SMILES string

O=[c]1c(c1c3ccccc3)c2ccccc2

InChI

1S/C15H10O/c16-15-13(11-7-3-1-4-8-11)14(15)12-9-5-2-6-10-12/h1-10H

InChI key

HCIBTBXNLVOFER-UHFFFAOYSA-N

Analysis Note

Assay (HPLC, area%): ≥ 98.0 % (a/a)
Melting range (lower value): ≥ 118 °C
Melting range (upper value): ≤ 122 °C
Identity (IR): passes test

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service