Skip to Content
Merck
All Photos(1)

Key Documents

440205-M

Millipore

LY 83583

A cell-permeable, competitive inhibitor of soluble guanylate cyclase (IC₅₀ = 2 µM).

Synonym(s):

LY 83583, 6-Anilino-5,8-quinolinequinone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H10N2O2
CAS Number:
Molecular Weight:
250.25
MDL number:
UNSPSC Code:
12352200

Quality Level

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

red-purple

solubility

ethanol: 3 mg/mL
acetone: soluble

storage temp.

2-8°C

SMILES string

N(c3ccccc3)C1=CC(=O)c2ncccc2C1=O

InChI

1S/C15H10N2O2/c18-13-9-12(17-10-5-2-1-3-6-10)15(19)11-7-4-8-16-14(11)13/h1-9,17H

InChI key

GXIJYWUWLNHKNW-UHFFFAOYSA-N

General description

A cell-permeable, competitive inhibitor of soluble guanylate cyclase (IC50 = 2 µM). Lowers the production of cGMP levels in a wide range of tissues by blocking intracellular Ca2+ release, with negligible effect on cAMP levels. Inhibits nitric oxide-induced smooth muscle relaxation. Shown to inhibit interleukin-1-induced cGMP accumulation in cultured rat aortic vascular smooth muscle cells. Also inhibits neutrophil chemotaxis induced by NO donors.

Biochem/physiol Actions

Primary Target
soluble guanylate cyclase
Target IC50: 2 µM against soluble guanylate cyclase

Warning

Toxicity: Harmful & Carcinogenic / Teratogenic (E)

Reconstitution

Following reconstitution, store in the refrigerator (4°C). Stock solutions are stable for up to 1 month at 4°C or up to 6 months at -20°C.

Other Notes

Adachi, R., et al. 2000. Int. J. Immunopharmacol. 22, 855.
Kawada, T., et al. 1994. Gen. Pharmacol. 25, 1361.
Beasley, D., et al. 1991. J. Clin. Invest. 87, 602.
Pandol, S.J., et al. 1990. J. Biol. Chem.265, 12846.
Light, D.B., et al. 1989. Science 243, 383.
Mulsch, A. 1988. J. Pharmacol. Exp. Ther.247, 283.
Fleisch, J.H., et al. 1984. J. Pharmacol. Exp. Ther. 229, 681.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service