Skip to Content
Merck
All Photos(1)

Key Documents

203734-M

Millipore

Brevetoxin PbTx-3, Ptychodiscus brevis

Lipid-soluble polyether marine toxin produced by the red tide dinoflagellate, Ptychodiscus brevis, found along the Gulf Coast of Florida.

Synonym(s):

Brevetoxin PbTx-3, Ptychodiscus brevis, Pumiliotoxin, Dihydrobrevetoxin B

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C50H72O14
CAS Number:
Molecular Weight:
897.10
MDL number:
UNSPSC Code:
12352200

Quality Level

Assay

>95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

solubility

ethanol: 10 mg/mL
DMSO: 30 mg/mL
acetonitrile: 30 mg/mL
chloroform: 30 mg/mL
methanol: 30 mg/mL
water: sparingly soluble

storage temp.

−20°C

SMILES string

O1C2(C(OC8(C(OC9C(OC%10C(OC%11(C(OC(CC%11O)CC(=C)CO)C%10)C)C9)\C=C/C8)C2)C)CC3OC4C(OC5C(OC6C(OC7C(OC(=O)C=C7C)C6)(C5)C)CC4C)CCC31C)C

InChI

1S/C50H72O14/c1-25(24-51)14-28-17-37(52)50(8)41(54-28)19-33-34(61-50)18-32-29(55-33)10-9-12-46(4)42(58-32)23-49(7)40(62-46)21-39-47(5,64-49)13-11-30-44(60-39)26(2)15-31-36(56-30)22-48(6)38(57-31)20-35-45(63-48)27(3)16-43(53)59-35/h9-10,16,26,28-42,44-45,51-52H,1,11-15,17-24H2,2-8H3/b10-9-

InChI key

BKMHDYJRAAJTAD-KTKRTIGZSA-N

General description

Lipid-soluble polyether marine toxin produced by the red tide dinoflagellate, Ptychodiscus brevis, found along the Gulf Coast of Florida. Voltage-dependent Na+ channel activator that causes contractile paralysis in animal models by binding to a unique site on these channels. The excitatory action of brevetoxins on nerve and muscle membranes is responsible for a wide spectrum of the toxic effects, including massive transmitter release from nerve endings, muscle fasciculations, and ventricular fibrillation. The toxin does not bind to either tetrodotoxin or aconitine/veratridine sites.

Biochem/physiol Actions

Primary Target
Voltage-dependent Na+ channel activator

Warning

Toxicity: Highly Toxic (H)

Reconstitution

Following reconstitution, aliquot, purge with inert gas and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C. Store stock solutions in glass vials; brevetoxin will stick to plastic. Use PTFE or silicon pipette tips.

Other Notes

Baden, D.G., et al. 1994. Nat. Toxins2, 212.
Rein, K., et al. 1994. J. Org. Chem.59, 2107.
Edwards, R.A., et al. 1992. Mol. Brain Res.14, 64.
Trainer, V.L. 1991. Mol. Pharmacol.40, 988.
Tsai, M.C., and Chen, M.L. 1991. Br. J. Pharmacol.103, 1126.
Trainer, V.L., et al. 1990. ACS Symposium Series418, 166.
Baden, D.G., et al. 1988. Toxicon26, 97.
Poli, M.A., et al. 1986. Mol. Pharmacol.30, 129.
Shimizu, Y., et al. 1986. J. Am. Chem. Soc.108, 514.
Baden, D.G., et al. 1982. Toxicon19, 455.
Catterfall, W.A., and Risk, M. 1981. Mol. Pharmacol.19, 345.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service