Skip to Content
Merck

1.06757

SAFC

Methyl 4-hydroxybenzoate

EMPROVE® ESSENTIAL, Ph. Eur., BP, JP, NF

Pharma Manufacturing

Synonym(s):

Methyl 4-hydroxybenzoate, Methylparaben, 4-Hydroxybenzoic acid methyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
4-(OH)C6H4(COOCH3)
CAS Number:
Molecular Weight:
152.15
MDL number:
UNSPSC Code:
12352108
EC Index Number:
202-785-7

Agency

BP
JP
NF
Ph. Eur.

Quality Level

vapor pressure

0.000005 hPa ( 20 °C)

product line

EMPROVE® ESSENTIAL

Assay

99.0-100.5% (HPLC)
99.0-102.0% (acidimetric)

form

solid

autoignition temp.

>600 °C

technique(s)

API processing | cocrystal formation: suitable

pH

5.8 (20 °C in H2O, saturated solution)

mp

125 °C

solubility

1.88 g/L

density

1.38 g/cm3 at 20 °C

application(s)

liquid formulation
ophthalmics
pharmaceutical
semi-solid formulation
solid formulation

storage temp.

15-25°C

SMILES string

O(C)C(=O)c1ccc(cc1)O

InChI

1S/C8H8O3/c1-11-8(10)6-2-4-7(9)5-3-6/h2-5,9H,1H3

InChI key

LXCFILQKKLGQFO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Finding the right excipient that matches your needs as well as regulatory demands can be a complicated challenge in formulation. With our application know-how and regulatory expertise, we support you in every step of development, scale-up, and production.
As part of our Emprove® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.

Application

Methyl 4-hydroxybenzoate has antimicrobial characteristics and is used as preservative in pharmaceutical formulations and cosmetics. It can be combined with other parabens like propyl 4-hydroxybenzoate or ethyl 4-hydroxybenzoate.

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

The product is not intended for use as a biocide under global biocide regulations, including but not limited to US EPA′s Federal Insecticide Fungicide and Rodenticide Act, European Biocidal Products Regulation, Canada’s Pest Management Regulatory Agency, Turkey’s Biocidal Products Regulation, Korea’s Consumer Chemical Products and Biocide Safety Management Act (K-BPR) and others.

Pictograms

Environment

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

334.4 °F

Flash Point(C)

168 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Find key considerations for the selection and use of benzalkonium chloride, benzyl alcohol, benzaldehyde, parabens, benzoic acid, and sodium benzoate as preservatives.

Find key considerations for the selection and use of benzalkonium chloride, benzyl alcohol, benzaldehyde, parabens, benzoic acid, and sodium benzoate as preservatives.

Find key considerations for the selection and use of benzalkonium chloride, benzyl alcohol, benzaldehyde, parabens, benzoic acid, and sodium benzoate as preservatives.

Find key considerations for the selection and use of benzalkonium chloride, benzyl alcohol, benzaldehyde, parabens, benzoic acid, and sodium benzoate as preservatives.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service