1.03557
Parteck® SI 450 (Sorbitol)
EMPROVE® ESSENTIAL, NF, FCC, JSFA
Pharma Manufacturing
Synonym(s):
Parteck® Sorbitol, D-Glucitol
About This Item
Recommended Products
Agency
JSFA
NF
Quality Level
reg. compliance
FCC
product line
EMPROVE® ESSENTIAL
form
coarse powder
autoignition temp.
420 °C
pH
6-7 (20 °C, 100 g/L in H2O)
mp
94-96 °C
transition temp
flash point >100 °C
solubility
2350 g/L
density
1.49 g/cm3 at 20 °C
application(s)
liquid formulation
pharmaceutical
solid formulation
storage temp.
2-25°C
SMILES string
OC(C(O)C(O)CO)C(O)CO
InChI
1S/C6H14O6/c7-1-3(9)5(11)6(12)4(10)2-8/h3-12H,1-2H2
InChI key
FBPFZTCFMRRESA-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
As part of our Emprove® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.
Application
Legal Information
replaced by
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service