Skip to Content
Merck
All Photos(1)

Key Documents

860615P

Avanti

Sphingosyl PE (d18:1)

Avanti Research - A Croda Brand 860615P, powder

Synonym(s):

D-erythro-sphingosyl phosphoethanolamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H43N2O5P
CAS Number:
Molecular Weight:
422.54
UNSPSC Code:
12352211
NACRES:
NA.25

form

powder

packaging

pkg of 1 × 5 mg (860615P-5mg)
pkg of 1 × 50 mg (860615P-50mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860615P

lipid type

sphingolipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@](/C=C/CCCCCCCCCCCCC)(O)[C@@]([H])(N)COP([O-])(OCC[NH3+])=O

InChI

1S/C20H43N2O5P/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-20(23)19(22)18-27-28(24,25)26-17-16-21/h14-15,19-20,23H,2-13,16-18,21-22H2,1H3,(H,24,25)/b15-14+/t19-,20+/m0/s1

InChI key

BPQSOCLDFWBDKN-PPQGUHKJSA-N

General description

Sphingosyl PE (d18:1), also known as D-erythro-sphingosyl phosphoethanolamine (PE), structurally comprises PE(d18:1/16:0) attached to a N-16:0-sphingosyl chain.

Packaging

5 mL Amber Glass Screw Cap Vial (860615P-50mg)
5 mL Amber Glass Screw Cap Vial (860615P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Handbook of lipid bilayers
Handbook of Inorganic Compounds (2013)
Anders Björkbom et al.
Biochimica et biophysica acta, 1798(3), 453-460 (2009-11-17)
Inositol phosphorylceramides (IPCs) are a class of anionic sphingolipids with a single inositol-phosphate head group coupled to ceramide. IPCs and more complex glycosylated IPCs have been identified in fungi, plants and protozoa but not in mammals. IPCs have also been

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service