Skip to Content
Merck
All Photos(1)

Key Documents

860488P

Avanti

Safingol

Avanti Research - A Croda Brand 860488P, powder

Synonym(s):

L-threo-dihydrosphingosine (d18:0) z

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H39NO2
CAS Number:
Molecular Weight:
301.51
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

form

powder

packaging

pkg of 1 × 5 mg (860488P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860488P

lipid type

sphingolipids
bioactive lipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

CCCCCCCCCCCCCCC[C@@](O)([H])[C@@]([H])(N)CO

InChI

1S/C18H39NO2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-18(21)17(19)16-20/h17-18,20-21H,2-16,19H2,1H3

InChI key

OTKJDMGTUTTYMP-UHFFFAOYSA-N

General description

Safingol is a synthetic L-threo-stereoisomer of endogenous (D-erythro-) sphinganine. It acts as an inhibitor of protein kinase C and sphingosine kinase in vitro. Safingol is involved in ceramide generation and induction of apoptosis. It is known to stimulate antitumor activity of cisplatin.

Application

Safingol has been used as a substituent of dihydrosphingosine (dhSph) to increase the cytotoxic response in N-(4-hydroxyphenyl)retinamide (4-HPR)-resistant cells.

Packaging

5 mL Amber Glass Screw Cap Vial (860488P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Safingol (L-threo-sphinganine) induces autophagy in solid tumor cells through inhibition of PKC and the PI3-kinase pathway
Coward J, et al.
Autophagy, 5(2), 184-193 (2009)
Andrej Kováčik et al.
Langmuir : the ACS journal of surfaces and colloids, 34(1), 521-529 (2017-12-13)
Ceramides (Cer) are key components of the skin permeability barrier. Sphingosine-based CerNS and dihydrosphingosine-based CerNdS (dihydroCer) have two chiral centers; however, the importance of the correct stereochemistry in the skin barrier Cer is unknown. We investigated the role of the
A phase I clinical study of safingol followed by cisplatin: promising activity in refractory adrenocortical cancer with novel pharmacology
Carvajal RD, et al.
Journal of Clinical Oncology, 24(18), 13044-13044 (2006)
Evaluation of bioactive sphingolipids in 4-HPR-resistant leukemia cells
Apraiz A, et al.
BMC Cancer, 11(1), 477-477 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service