Skip to Content
Merck
All Photos(1)

Key Documents

860385P

Avanti

16:0-18:1 D5 PG

1-hexadecanoyl-2-(9Z-octadecenoyl)-sn-glycero-3-phospho-(1′-rac-glycerol-1′,1′,2′,3′,3′-d5) (ammonium salt), powder

Synonym(s):

1-palmitoyl-2-oleoyl-sn-glycero-3-phospho-(1′racglycerol-d5) (ammonium salt); 110899

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C40H75D5NO10P
CAS Number:
Molecular Weight:
771.07
UNSPSC Code:
12352100
NACRES:
NA.25

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (860385P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860385P

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OC([2H])([2H])C(O)([2H])C([2H])([2H])O)=O)(OC(CCCCCCC/C=C\CCCCCCCC)=O)COC(CCCCCCCCCCCCCCC)=O.[NH4+]

InChI key

WFYVYFVKHCDATR-GKRQAWATSA-N

Application

1-hexadecanoyl-2-(9Z-octadecenoyl)-sn-glycero-3-phospho-(1′-rac-glycerol-1′,1′,2′,3′,3′-d5) (16:0-18:1 D5 PG) has been used as a deuterated standard to test the ion suppression of sea water samples by high performance liquid chromatography (HPLC) analysis. It has also been used as an internal standard to verify fragmentation pattern and elution times of different lipid class for lipid identification.

Packaging

5 mL Clear Glass Sealed Ampule (860385P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Melissa S Roth et al.
The Plant cell, 31(3), 579-601 (2019-02-23)
Light and nutrients are critical regulators of photosynthesis and metabolism in plants and algae. Many algae have the metabolic flexibility to grow photoautotrophically, heterotrophically, or mixotrophically. Here, we describe reversible Glc-dependent repression/activation of oxygenic photosynthesis in the unicellular green alga
Kevin W Becker et al.
Nature communications, 9(1), 5179-5179 (2018-12-07)
Sunlight is the dominant control on phytoplankton biosynthetic activity, and darkness deprives them of their primary external energy source. Changes in the biochemical composition of phytoplankton communities over diel light cycles and attendant consequences for carbon and energy flux in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service