Skip to Content
Merck
All Photos(2)

Key Documents

855800P

Avanti

24:0 Lyso PC

1-lignoceroyl-2-hydroxy-sn-glycero-3-phosphocholine, powder

Synonym(s):

1-tetracosanoyl-sn-glycero-3-phosphocholine; PC(24:0/0:0)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H66NO7P
CAS Number:
Molecular Weight:
607.84
MDL number:
UNSPSC Code:
51191904
NACRES:
NA.25

Assay

>99% (LPC; may contain up to 10% of the 2-LPC isomer, TLC)

form

powder

packaging

pkg of 1 × 25 mg (855800P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 855800P

shipped in

dry ice

storage temp.

−20°C

Application

24:0 Lyso PC has been used as a standard for liquid chromatography with tandem mass spectrometry (LC-MS/MS) analysis.

Packaging

5 mL Amber Glass Screw Cap Vial (855800P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

also commonly purchased with this product

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The peroxisomal AAA ATPase complex prevents pexophagy and development of peroxisome biogenesis disorders
Law KB, et al.
Autophagy, 13(5), 868-884 (2017)
Kelsey B Law et al.
Autophagy, 13(5), 868-884 (2017-05-20)
Peroxisome biogenesis disorders (PBDs) are metabolic disorders caused by the loss of peroxisomes. The majority of PBDs result from mutation in one of 3 genes that encode for the peroxisomal AAA ATPase complex (AAA-complex) required for cycling PEX5 for peroxisomal

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service