Key Documents
SYNV115712
Synthware dual vacuum trap assembly, complete
with #15 o-ring joints
Synonym(s):
Synthware V115712
About This Item
UNSPSC Code:
40161501
NACRES:
NB.51
Pricing and availability is not currently available.
manufacturer/tradename
V115712
General description
Synthware dual vacuum trap complete assembly includes O-rings and clamps. It is designed for use with manifolds having #15 O-ring joints and features 0-12 mm high vacuum valves as the shut-off. The pre-trap utilizes two 28/15 ball joints to add flexibility to the assembly. Additionally, the pre-trap uses a 57 mm horseshoe clamp and two 57 mm O-ring flanges for quick connection.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)