SYNF761425
Synthware™ storage tube with standard taper side arm
capacity 25 mL, joint: ST/NS 14/20, valve size 0-4 mm
Synonym(s):
Synthware F761425, storage tube with outer joint
All Photos(1)
About This Item
UNSPSC Code:
41121800
NACRES:
NB.51
Recommended Products
material
black cap
cap (plug seal)
colorless tube
glass tube
round bottom
Joint
joint: ST/NS14/20
packaging
pkg of 1 ea
manufacturer/tradename
Synthware F761425
capacity
25 mL
valve size
0-4 mm
Related Categories
General description
Supplied with high vacuum valve with PTFE O-ring at tip.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)