Key Documents
SYNA842415
Synthware vacuum filtration adapter
bottom joint: ST/NS 24/40, Stopper Size: #3, joint: ST/NS 24/40, stopper size #3
Synonym(s):
Synthware A842415, Glass adapter vacuum filtration
About This Item
UNSPSC Code:
41121815
NACRES:
NB.51
Pricing and availability is not currently available.
Joint
bottom joint: ST/NS24/40
joint: ST/NS24/40
packaging
pkg of 1 ea
manufacturer/tradename
A842415
Synthware A842415
Related Categories
General description
Used with plain stem Buchner funnels and pluro-stopper. Hose connection has an O.D of 10 mm at the largest serration. Order pluro-stoppers below separately.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)