Key Documents
MT51344724
Mettler-Toledo perfectION combination electrode
About This Item
UNSPSC Code:
39121436
NACRES:
NB.79
material
glass membrane (Na+)
glass shaft
description
Reference electrolyte 3 mol/L KCl
S7 connection
Without cable
manufacturer/tradename
Mettler-Toledo 51344724
temperature measuring range
0-80 °C
pH range
8-11
General description
Mettler Toledo perfectION ISE Combined ion-selective electrode means ISE measurement has never been so simple!
The new Click & Clear junction of the combined ion-selective electrodes combines three advantages in one tool: The electrolyte solution can flow easily through the junction by pressing the electrode head downwards. This ensures that the junction is optimally cleaned between the measurements and the electrolyte can be simply emptied and refilled if necessary.
Click & Clear is a sleeve junction that enables optimum contact between the electrolyte and measurement solution, even in difficult samples.
Features and Benefits
All combined ion-selective electrodes consist of a measuring electrode complete with an integrated reference. There is therefore no need for an additional reference electrode.
  • Sensor with attached cable and BNC connector
  • Reference electrolyte (60 mL)
  • Operating instructions
    To ensure an unproblematic start to your ISE measurement, the required calibration standards and ISA solutions (Ionic Strength Adjuster) can be ordered at the same time.
comb Na
Legal Information
Click & Clear is a trademark of Mettler-Toledo, Inc.
perfectION is a trademark of Mettler-Toledo, Inc.
related product
Choose from one of the most recent versions:
Certificates of Analysis (COA)