Skip to Content
Merck
All Photos(2)

Key Documents

D44405

Sigma-Aldrich

α,α′-Dibromo-o-xylene

97%

Synonym(s):

1,2-Bis(bromomethyl)benzene, o-Xylylene dibromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4(CH2Br)2
CAS Number:
Molecular Weight:
263.96
Beilstein:
637159
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

91-94 °C (lit.)

density

1.96 g/mL at 25 °C (lit.)

SMILES string

BrCc1ccccc1CBr

InChI

1S/C8H8Br2/c9-5-7-3-1-2-4-8(7)6-10/h1-4H,5-6H2

InChI key

KGKAYWMGPDWLQZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service