Skip to Content
Merck
All Photos(1)

Key Documents

BR781668

BRAND® microplate BRANDplates®, pureGrade

size 96 wells, size 350 μL, flat bottom, black

Synonym(s):

BRANDplates®, Microplates, pureGradeS

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.22
material:
black wells
flat bottom
polystyrene
size:
350 μL
96 wells
working volume:
350 μL
Pricing and availability is not currently available.

material

black wells
flat bottom
polystyrene

sterility

sterile

feature

lid
skirt (F-bottom)

packaging

pack of 50 ea
pkg of (individually packaged)

manufacturer/tradename

BRAND 781668

size

350 μL
96 wells

working volume

350 μL

color

black

binding type

non-treated surface

Looking for similar products? Visit Product Comparison Guide

General description

The BRAND® Microplate BRANDplates® PureGrade features a non-treated surface, making it particularly well-suited for bacteriological assays. Sterile according to ISO 11137 standards, this microplate ensures a contamination-free environment for sensitive experiments. It is classified as BIO-CERT® CELL CULTURE QUALITY, guaranteeing that it is free from human DNA, RNase, DNase, and pyrogens, and is non-cytotoxic. This reliable microplate is an essential tool for researchers conducting microbiological studies, providing consistent performance and safety in cell culture applications.

Features and Benefits

• Optimal surfaces for optimal results• Reliable quality from the cleanroom• Tested by independent laboratories

Legal Information

BIO-CERT is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
BRANDplates is a registered trademark of BRAND GMBH + CO KG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service