BR781622
BRAND® microplate BRANDplates®, pureGrade
size 384 wells, size 100 μL, flat bottom, black
Synonym(s):
BRANDplates®, Microplates, medium binding, pureGrade
About This Item
flat bottom
polystyrene
384 wells
Recommended Products
material
black wells
flat bottom
polystyrene
sterility
non-sterile
feature
lid: no
skirt Skirt (F-bottom)
packaging
pack of 50 ea
manufacturer/tradename
BRAND 781622
size
100 μL
384 wells
working volume
100 μL
color
black
suitability
suitable for (bacteriological assays)
binding type
medium binding surface
Looking for similar products? Visit Product Comparison Guide
General description
Features and Benefits
- Optimal surfaces for optimal results
- Reliable quality from the cleanroom
- Tested by independent laboratories
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service