Skip to Content
Merck
All Photos(1)

Key Documents

901187

Sigma-Aldrich

Molybdenum disulfide ink for inkjet printing

greener alternative

Synonym(s):

Molybdenum disulfide dispersion, with ethyl cellulose in cyclohexanone and terpineol

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352103
NACRES:
NA.23

description

Resistivity: 10-100 kΩcm (thermally annealed 400 °C for 3 hours in Ar/H2 environment, film thickness >150 nm)

Quality Level

form

dispersion

composition

Solid content, 3.0-3.85%

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

color

green

particle size

≤0.4 μm

surface tension

30-35 dyn/cm

viscosity

4-11 mPa.s(25 °C) (shear viscoisty at 1000 s-1, 25 °C)

bp

213-218 °C (Terpineol)

density

0.9-1.1 g/mL at 25 °C

greener alternative category

SMILES string

[Mo]=S=S

InChI

1S/Mo.S2/c;1-2

InChI key

IZCXEXNUAYYFRY-UHFFFAOYSA-N

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product belongs to Enabling category of greener alternatives thus aligns with "Design for energy efficency". Click here for more information.

Legal Information

Sold under Materials Transfer Agreement with Mark Hersam group at Northwestern University.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

114.8 °F - closed cup

Flash Point(C)

46 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Recent advancements in paper-based sensing platforms offer cost-effective clinical diagnostics with microfluidic channels and colorimetric or electrochemical detection zones.

Recent advancements in paper-based sensing platforms offer cost-effective clinical diagnostics with microfluidic channels and colorimetric or electrochemical detection zones.

Recent advancements in paper-based sensing platforms offer cost-effective clinical diagnostics with microfluidic channels and colorimetric or electrochemical detection zones.

Recent advancements in paper-based sensing platforms offer cost-effective clinical diagnostics with microfluidic channels and colorimetric or electrochemical detection zones.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service