Skip to Content
Merck
All Photos(1)

Key Documents

492450

Sigma-Aldrich

Mevalonolactone-1,2-13C2

99 atom % 13C, 98% (CP)

Synonym(s):

DL-Mevalonic-1,2-13C2 lactone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13C2C4H10O3
CAS Number:
Molecular Weight:
132.13
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

99 atom % 13C

Assay

98% (CP)

form

solid

refractive index

n20/D 1.473 (lit.)

bp

150 °C/5 mmHg (lit.)

mp

28 °C (lit.)

mass shift

M+5

storage temp.

2-8°C

SMILES string

[2H]C([2H])([2H])C1(O)CCO[13C](=O)[13CH2]1

InChI

1S/C6H10O3/c1-6(8)2-3-9-5(7)4-6/h8H,2-4H2,1H3/i4+1,5+1

InChI key

JYVXNLLUYHCIIH-MQIHXRCWSA-N

Application

This product may be used as an analytical standard.

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F

Flash Point(C)

113 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service