Skip to Content
Merck
All Photos(1)

Key Documents

141674

Sigma-Aldrich

1-n-Propylpiperazine dihydrobromide

96%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H16N2 · 2HBr
CAS Number:
Molecular Weight:
290.04
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

96%

mp

259.5-264.5 °C (lit.)

solubility

water: soluble 50 mg/mL, clear, colorless

SMILES string

Br.Br.CCCN1CCNCC1

InChI

1S/C7H16N2.2BrH/c1-2-5-9-6-3-8-4-7-9;;/h8H,2-7H2,1H3;2*1H

InChI key

YTPQLWVHCBATKO-UHFFFAOYSA-N

General description

1-n-Propylpiperazine dihydrobromide reacts with potassium thiocyanate to yield 1-(4-n-propyl)piperazine thioamide.

Application

1-n-Propylpiperazine dihydrobromide was used in the synthesis of amidines and sulfonamides of 5-and 6-amino-2,3-bis(4-alkyl-1-piperazazinyl)quinoxalines.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anna Frymarkiewicz et al.
European journal of medicinal chemistry, 44(4), 1674-1681 (2008-11-08)
A series of 1-[[2-thiazol-5-yl-(2-aminoethyl)]-4-n-propyl]piperazine derivatives have been prepared and in vitro tested as H(3)-receptor antagonists (the electrically evoked contraction of the guinea pig jejunum). It appeared that by comparison of homologous pairs the 1-[[2-thiazol-5-yl-(2-methyl-2-phenylalkylaminoethyl)]-4-n-propyl]piperazine derivatives (4c1-4c3) have slightly higher activity
P F Fabio et al.
Journal of medicinal chemistry, 23(2), 201-206 (1980-02-01)
A series of amidines and sulfonamides of 5- and 6-amino-2,3-bis(4-alkyl-1-piperazinyl)quinoxalines was synthesized and tested against cecal and hepatic forms of Entamoeba histolytica infections in rats and hamsters, respectively. Four compounds (5, 6, 8, and 9) were found to have acceptable

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service