Skip to Content
Merck
All Photos(2)

Key Documents

113409

Sigma-Aldrich

5-Methyl-2-nitrobenzoic acid

95%

Synonym(s):

6-Nitro-m-toluic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H3(NO2)CO2H
CAS Number:
Molecular Weight:
181.15
Beilstein:
1959031
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

solid

mp

134-136 °C (lit.)

functional group

carboxylic acid
nitro

SMILES string

Cc1ccc(c(c1)C(O)=O)[N+]([O-])=O

InChI

1S/C8H7NO4/c1-5-2-3-7(9(12)13)6(4-5)8(10)11/h2-4H,1H3,(H,10,11)

InChI key

QRRSIFNWHCKMSW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

5-Methyl-2-nitrobenzoic acid is used in the preparation of methyl-N-(5-methyl-2-nitrophenyl) carbamate.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

New preparation of benzo-2, 1, 3-oxadiazoles (benzofurazans).
Prokipcak JM and Forte PA.
Canadian Journal of Chemistry, 48(19), 3059-3063 (1970)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service