Merck
  • Effects of atrazine, endosulfan and butylated hydroxyanisole on glutathione-S-transferases in Orthosia gothica.
Effects of atrazine, endosulfan and butylated hydroxyanisole on glutathione-S-transferases in Orthosia gothica.
Comparative biochemistry and physiology. Toxicology & pharmacology : CBP (2000-11-18)
E Egaas
ABSTRACT
Atrazine (1,000 ppm), endosulfan (1 ppm) or butylated hydroxyanisole (BHA) (1,000 ppm) added to a semi-synthetic diet of Orthosia gothica for 2 days in the last instar did not change the soft tissue cytosolic glutathione-S-transferase (GST) activities towards 1-chloro-2,4-dinitrobenzene (CDNB), 1,2-dichloro-4-nitrobenzene (DCNB) and cumene hydroperoxide (CU). However, all three pesticides changed the GST subunit composition compared with the control as observed by reverse phase high performance liquid chromatography of the isozymes purified by glutathione-Sepharose affinity chromatography. The changes seem to have occurred mainly in the GST class 2 subunit. There is no obvious explanation for the changes, which may be a result of interactions between xenobiotic and GST in the cytosol as well as changes in the level of regulation of synthesis. However, the observation added to our knowledge of the processes involved when pesticides are degraded by GSTs in vivo.
Sigma-Aldrich
1,2-Dichloro-4-nitrobenzene, 99%
Pricing and availability is not currently available.