Skip to Content
Merck

Skip To

1073423

USP

Biphenyl

United States Pharmacopeia (USP) Reference Standard

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Linear Formula:
C6H5C6H5
CAS Number:
Molecular Weight:
154.21
UNSPSC Code:
41116107
NACRES:
NA.24
PubChem Substance ID:
MDL number:
Beilstein/REAXYS Number:
1634058
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

vapor density

5.31 (vs air)

vapor pressure

9.46 mmHg ( 115 °C)

API family

fenbufen

autoignition temp.

1004 °F

expl. lim.

0.6 %, 111 °F, 5.8 %, 166 °F

manufacturer/tradename

USP

bp

255 °C (lit.)

mp

68-70 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

c1ccc(cc1)-c2ccccc2

InChI

1S/C12H10/c1-3-7-11(8-4-1)12-9-5-2-6-10-12/h1-10H

InChI key

ZUOUZKKEUPVFJK-UHFFFAOYSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Biphenyl USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
PHR235835800B34656
manufacturer/tradename

USP

manufacturer/tradename

-

manufacturer/tradename

-

manufacturer/tradename

-

grade

pharmaceutical primary standard

grade

certified reference material, pharmaceutical secondary standard

grade

analytical standard

grade

-

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical small molecule

application(s)

agriculture
environmental

application(s)

-

format

neat

format

-

format

neat

format

-

vapor pressure

9.46 mmHg ( 115 °C)

vapor pressure

9.46 mmHg ( 115 °C)

vapor pressure

9.46 mmHg ( 115 °C)

vapor pressure

9.46 mmHg ( 115 °C)

expl. lim.

0.6 %, 111 °F, 5.8 %, 166 °F

expl. lim.

0.6 %, 111 °F, 5.8 %, 166 °F

expl. lim.

0.6 %, 111 °F, 5.8 %, 166 °F

expl. lim.

0.6 %, 111 °F, 5.8 %, 166 °F


Still not finding the right product?

Explore all of our products under Biphenyl


pictograms

Exclamation markEnvironment

signalword

Warning

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

13 - Non Combustible Solids

wgk

WGK 2

flash_point_f

230.0 °F - closed cup

flash_point_c

110 °C - closed cup



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters