Skip to Content
Merck
All Photos(13)

Key Documents

ZRB2112

Sigma-Aldrich

Anti-CD44 Antibody, clone 5H23 Antibody, ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

CD44 antigen;CDw44;Epican;Extracellular matrix receptor III;ECMR-III;GP90 lymphocyte homing/adhesion receptor;HUTCH-I;Heparan sulfate proteoglycan;Hermes antigen;Hyaluronate receptor;Phagocytic glycoprotein 1;PGP-1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

5H23, recombinant monoclonal

description

5H23 Clone

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 81.54 kDa
observed mol wt ~140 kDa

purified by

using Protein A

species reactivity

human

species reactivity (predicted by homology)

monkey

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

ELISA: suitable
affinity binding assay: suitable
flow cytometry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

C-terminal half

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... CD44(960)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 5H23 is a ZooMAb® rabbit recombinant monoclonal antibody that specifically detects CD44. It targets an epitope within 16 amino acids from the extracellular domain within the C-terminal half.

Immunogen

KLH-conjugated linear peptide corresponding to 16 amino acids from the C-terminal half, extracellular domain of human CD44.

Application

Quality Control Testing

Evaluated by Western Blotting with recombinant Human CD44.

Western Blotting Analysis (WB): A 1:10,000 dilution of this antibody detected recombinant Human CD44.

Tested Applications

Enzyme Immunoassay Analysis: A Series of Dilutions from a representative lot detected CD44 in CD44 and GST proteins.

Flow Cytometry Analysis: 1 μg from a representative lot detected CD44 in one million Human peripheral blood mononuclear cells (PBMC).

Immunohistochemistry (Paraffin) Analysis: A 1:1,000 dilution from a representative lot detected CD44 in human skin tissue sections.

Affinity Binding Assay: A representative lot of this antibody bound recombinant Human CD44 protein with a KD of 2.2 x 10-6 in an affinity binding assay.

Immunofluorescence Analysis: A 1:1,000 dilution from a representative lot detected CD44 in Human skin tissue sections.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

CD44 antigen (UniProt: P16070; also known as CDw44, Epican, Extracellular matrix receptor III, ECMR-III, GP90 lymphocyte homing/adhesion receptor, HUTCH-I, Heparan sulfate proteoglycan, Hermes antigen, Hyaluronate receptor, Phagocytic glycoprotein 1, PGP-1, CD44) is encoded by the CD44 (also known as LHR, MDU2, MDU3, MIC4) gene (Gene ID: 960) in human. CD44 is a single-pass type I, highly glycosylated membrane protein that is synthesized with a signal peptide (aa 1-20), which is subsequently cleaved off to generate the mature form that contains an extracellular domain (aa 21-649), a transmembrane domain (aa 650-670), and a cytoplasmic domain (aa 671-742). Nineteen different isoforms of CD44 have been described that are produced by alternative splicing. All isoforms possess a hyaluronic acid (HA)-binding domain in its N-terminal region of the extracellular domain. CD44 expression is observed on all leukocytes, endothelial cells, hepatocytes, and mesenchymal cells. It binds to several ligands including hyaluronic acid (HA), Osteopontin, chondroitin, collagen, fibronectin, and serglycin/sulfated proteoglycan. However, the main ligand for CD44 is HA that is expressed by stromal and cancer cells. HA binds the CD44 ligand binding domain and induces conformational changes that allow binding of adaptor proteins or cytoskeletal elements to intracellular domains. This leads to the activation of various signaling pathways leading to cell proliferation, adhesion, migration, and invasion. Overexpression of CD44 has been reported in several cell types and it is considered as an ideal marker for cancer stem cells. Its interaction with appropriate extracellular matrix ligands promotes the migration and invasion processes involved in tumor metastases. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Chen, C., et al. (2018). J. Hematol. Oncol. 11; Article 64). Senbanio, LT., and Chellaiah, MA (2017). Front. Cell Dev. Biol. 5; 18).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

300 μg/mL after reconstitution at 25 μL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service