Skip to Content
Merck
All Photos(9)

Key Documents

ZRB1367

Sigma-Aldrich

Anti-C-EBP alpha Antibody, clone 2E17, ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

C/EBP alpha, CCAAT/enhancer-binding protein alpha

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit (recombinant)

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

2E17, monoclonal
recombinant monoclonal

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 37.56 kDa
observed mol wt ~50 kDa

species reactivity

human, mouse

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

flow cytometry: suitable
immunocytochemistry: suitable
western blot: suitable

isotype

IgG

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... CEBPA(1050)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb antibodies represent an entirely new generation of recombinant monoclonal antibodies.

Each ZooMAb antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb antibodies are reliably available and ready to ship when you need them.

Learn more about ZooMAb here.

Specificity

Clone 2E17 is a ZooMAb rabbit recombinant monoclonal antibody that specifically detects C-EBP alpha. It targets an epitope within 147 amino acids from the N-terminal region.

Immunogen

GST/His-tagged recombinant fragment corresponding to 147 amino acids from N-terminal region of human C-EBP alpha, isoform 1.

Application

Anti-C-EBP alpha, clone 2E17 ZooMAb, Cat. No. ZRB1367, a recombinant Rabbit monoclonal antibody that specifically targets C-EBP alpha and has been tested for use in Flow Cytometry, Immunocytochemistry, and Western Blotting.
Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected C-EBP alpha in THP-1 cell lysate.

Flow Cytometry Analysis: 0.01 μg from a representative lot detected C-EBP alpha in one million U937 cells.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected C-EBP alpha in THP-1 cells.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

CCAAT/enhancer-binding protein alpha (UniProt: P49715; also known as C/EBP alpha, C-EBP alpha) is encoded by the CEBPA (also known as CEBP) gene in human. C-EBP alpha belong to the bZIP family of proteins that serves as a transcription factor that coordinates proliferation arrest and the differentiation of myeloid progenitors, adipocytes, hepatocytes, and cells of the lung and the placenta. It binds directly to the consensus DNA sequence 5′-T[TG]NNGNAA[TG]-3′ acting as an activator on distinct target genes. It can bind DNA as a homodimer or as a stable heterodimer with CEBPB, CEBPD, CEBPE, and CEBPG. It is reported to be essential for the transition from common myeloid progenitors (CMP) to granulocyte/monocyte progenitors (GMP) and has also been shown to be critical for the proper development of the liver and the lung and for terminal adipocyte differentiation. C-EBP alpha down-regulates the expression of genes that maintain cells in an undifferentiated and proliferative state through E2F1 repression, which is critical for its ability to induce adipocyte and granulocyte terminal differentiation. It can be phosphorylated on multiple sites. Phosphorylation at serine 190 is shown to be essential for its interaction with CDK2, CDK4 and SWI/SNF complex leading to cell cycle inhibition. Phosphorylation at threonine 226 and threonine 230 by GSK3 is constitutive in adipose and lung tissue and these sites are also shown to be phosphorylated in liver during feeding, but not during fasting cycle. Four isoforms of C-EBP alpha have been described that are produced by alternative splicing. Mutations in CEBPA gene are known to cause acute myelogenous leukemia (AML). This ZooMAb recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Wedel, A., et al. (1995). Immunobiology. 193(2-4); 171-185; Johnson, PF et al. (2005). J. Cell Sci. 118(12); 2545-2555).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service