Skip to Content
Merck

Z710784

Tunair shake flask

flask volume 300 mL, polypropylene flask, half baffle

Synonym(s):

baffled flask, baffled flasks

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NB.22
UNSPSC Code:
41122104
Binding type:
non-treated surface
Material:
polypropylene flask, straight neck, wide-mouth
Sterility:
non-sterile
Packaging:
pk of 1 ea, case of 4 ea
Feature:
without cap
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

polypropylene flask, straight neck, wide-mouth

sterility

non-sterile

feature

without cap

packaging

pk of 1 ea, case of 4 ea

manufacturer/tradename

IBI SS-4002

flask volume

300 mL

working volume

100 mL

binding type

non-treated surface

General description

Autoclavable Plastics - Contact clear polypropylene
Working Volumes - 100 mL for 300 mL flasks (150-200 rpm for 2 in. throw)
High 02 absorption rate achievable by unique baffle and turbo-vane closure design. CAP-FLO closure with DRI-GAUZE filter lining provides uniformity and consistency in gas diffusion.
Flasks, closures, filters or membrane linings are sterilizable by autoclaving. Wide-mouth opening allowing easy access for media preparation, harvest and cleanup. Barrel-shaped design provides wide vortex and more operational capacity than conventional Erlenmeyer flasks. Slip-on closure protects flask neck from airborne particles and eliminates the need for flaming. Provides better culture growth and productivity than Erlenmeyer flask.
Caps are not included with flasks.
  • CAP-SIL closure with silicone membrane lining minimizes evaporation.
  • 300 mL flasks for front-line culture development studies.
  • 2.5L flask for production studies, equipped with a handle.

Legal Information

CAP-FLO is a trademark of IBI Scientific
CAP-SIL is a trademark of IBI Scientific
DRI-GAUZE is a trademark of IBI Scientific
Tunair is a trademark of IBI Scientific


Still not finding the right product?

Explore all of our products under Tunair shake flask




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library