Skip to Content
Merck
All Photos(1)

Key Documents

TMO337516

Nunc® cryogenic tubes

polypropylene, capacity 4.5 mL, round bottom, starfoot, external thread, sterile, case of 1200 ea

Synonym(s):

biobanking tube, cell culture cryogenic tube

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121704
NACRES:
NB.12

material

polypropylene
round bottom

sterility

sterile

packaging

case of 1200 ea

manufacturer/tradename

Nunc 337516

technique(s)

cryopreservation: suitable

capacity

4.5 mL

Looking for similar products? Visit Product Comparison Guide

General description

Nunc®cryogenic tubes are designed for the storage of samples, ranging from general cold storage to the vapor phase of liquid nitrogen. These tubes are available with either internal or external threads to suit different preferences and requirements.

Legal Information

Nunc is a registered trademark of Thermo Fisher Scientific or its subsidiaries

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service