Skip to Content
Merck
All Photos(1)

Key Documents

SML3523

Sigma-Aldrich

Solriamfetol hydrochloride

≥98% (HPLC)

Synonym(s):

(2R)-2-Amino-3-phenylpropyl carbamate hydrochloride, (R)-2-amino-3-phenylpropylcarbamate hydrochloride, ADX-N 05, ADX-N05, JZP 110, JZP-110

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H14N2O2 ·HCl
CAS Number:
Molecular Weight:
230.69
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.04

Quality Level

Assay

≥98% (HPLC)

form

powder

drug control

USDEA Schedule IV

storage condition

desiccated

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

Biochem/physiol Actions

Solriamfetol (JZP-110) is a potent and selective dual reuptake inhibitor at dopamine and norepinephrine transporters that increases extracellular levels of dopamine and norepinephrine in vivo. Solriamfetol is a wake-promoting agent.

Caution

Hygroscopic

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Richard K Bogan et al.
Sleep medicine, 16(9), 1102-1108 (2015-08-25)
JZP-110 is a wake-promoting agent with dopaminergic and noradrenergic activity. This double-blind, crossover study, randomized adults with narcolepsy with or without cataplexy (N = 33) to placebo or JZP-110 at 150 mg/day (weeks 1 and 3) increased to 300 mg/day (weeks 2 and 4).
Michelle G Baladi et al.
The Journal of pharmacology and experimental therapeutics, 366(2), 367-376 (2018-06-13)
Excessive sleepiness (ES) is associated with several sleep disorders, including narcolepsy and obstructive sleep apnea (OSA). A role for monoaminergic systems in treating these conditions is highlighted by the clinical use of US Food and Drug Administration-approved drugs that act
Stefano de Biase et al.
Expert opinion on investigational drugs, 26(8), 953-963 (2017-07-21)
Narcolepsy is a chronic sleep disorder characterized by a pentad of excessive daytime sleepiness (EDS), cataplexy, sleep paralysis, hypnagogic/hypnopompic hallucinations, and disturbed nocturnal sleep. While non-pharmacological treatments are sometimes helpful, more than 90% of narcoleptic patients require a pharmacological treatment.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service