Skip to Content
Merck

SMB00522

5,6,7-Trihydroxy-4′-methoxyflavone

≥85% (LC/MS-UV)

Synonym(s):

5,6,7-trihydroxy-2-(4-methoxyphenyl)chromen-4-one

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C16H12O6
CAS Number:
Molecular Weight:
300.26
NACRES:
NA.25
PubChem Substance ID:
UNSPSC Code:
12352205
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥85% (LC/MS-UV)

form

solid

solubility

DMSO: 1 mg/mL

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

O=C1C2=C(C(O)=C(C=C2OC(C3=CC=C(OC)C=C3)=C1)O)O

InChI

1S/C16H12O6/c1-21-9-4-2-8(3-5-9)12-6-10(17)14-13(22-12)7-11(18)15(19)16(14)20/h2-7,18-20H,1H3

InChI key

XVMMEYCPXZYLAI-UHFFFAOYSA-N

General description

Natural prodcut derived from plant source.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library