Skip to Content
Merck
All Photos(1)

Key Documents

SAB1408054

Sigma-Aldrich

Anti-RNF146 antibody produced in mouse

purified immunoglobulin, buffered aqueous solution

Synonym(s):

DKFZp434O1427, RP3-351K20.1, dJ351K20.1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
conjugate:
unconjugated
application:
WB
clone:
polyclonal
species reactivity:
human
citations:
5
technique(s):
western blot: 1 μg/mL

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~38.8 kDa

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RNF146(81847)

Immunogen

RNF146 (NP_112225.2, 1 a.a. ~ 358 a.a) full-length human protein.

Sequence
MAGCGEIDHSINMLPTNRKANESCSNTAPSLTVPECAICLQTCVHPVSLPCKHVFCYLCVKGASWLGKRCALCRQEIPEDFLDKPTLLSPEELKAASRGNGEYAWYYEGRNGWWQYDERTSRELEDAFSKGKKNTEMLIAGFLYVADLENMVQYRRNEHGRRRKIKRDIIDIPKKGVAGLRLDCDANTVNLARESSADGADSVSAQSGASVQPLVSSVRPLTSVDGQLTSPATPSPDASTSLEDSFAHLQLSGDNTAERSHRGEGEEDHESPSSGRVPAPDTSIEETESDASSDSEDVSAVVAQHSLTQQRLLVSNANQTVPDRSDRSGTDRSVAGGGTVSVSVRSRRPDGQCTVTEV

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

RANKL coordinates multiple osteoclastogenic pathways by regulating expression of ubiquitin ligase RNF146.
Matsumoto Y, et. al.
The Journal of Clinical Investigation, 127, 1303-1303 (2017)
Ubiquitin ligase RNF146 coordinates bone dynamics and energy metabolism.
Matsumoto Y, et. al.
The Journal of Clinical Investigation, 127, 2612-2612 (2017)
Yoshinori Matsumoto et al.
The Journal of clinical investigation, 127(7), 2612-2625 (2017-06-06)
Cleidocranial dysplasia (CCD) is an autosomal dominant human disorder characterized by abnormal bone development that is mainly due to defective intramembranous bone formation by osteoblasts. Here, we describe a mouse strain lacking the E3 ubiquitin ligase RNF146 that shows phenotypic
Yoshinori Matsumoto et al.
The Journal of clinical investigation, 127(4), 1303-1315 (2017-03-14)
Bone undergoes continuous remodeling due to balanced bone formation and resorption mediated by osteoblasts and osteoclasts, respectively. Osteoclasts arise from the macrophage lineage, and their differentiation is dependent on RANKL, a member of the TNF family of cytokines. Here, we
Arun A Chandrakumar et al.
The Journal of cell biology, 220(7) (2021-06-16)
Rab11 GTPase proteins are required for cytokinesis, ciliogenesis, and lumenogenesis. Rab11a is critical for apical delivery of podocalyxin (PODXL) during lumen formation in epithelial cells. SH3BP5 and SH3BP5L are guanine nucleotide exchange factors (GEFs) for Rab11. We show that SH3BP5

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service