Skip to Content
Merck

MTCC2410

MTC Bio Screw Cap Centrifuge Tube

capacity 10 mL, conical bottom, sterile, pack of × 1000 ea 20 bags of 50ea

Synonym(s):

conical tube, plastic tube, MTC Bio C2410, sample collection tube, sample receptacle, sample tube

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NB.78
UNSPSC Code:
41121703
Capacity:
10 mL
Material:
blue cap, clear tube, conical bottom , polyethylene cap, polypropylene , screw cap
Sterility:
sterile
Packaging:
pack of × 1000 ea 20 bags of 50ea
Parameter:
12,000 × g speed
Feature:
graduation
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

blue cap, clear tube, conical bottom , polyethylene cap, polypropylene , screw cap

sterility

sterile

feature

graduation

packaging

pack of × 1000 ea 20 bags of 50ea

manufacturer/tradename

MTC Bio C2410

parameter

12,000 × g speed

capacity

10 mL

diam. × L

16 mm × 104 mm

General description

Centrifuge Tube, 10mL, PP, sterile, flat screw cap, 20 bags of 50 tubes, 1000/cs
These centrifuge tubes are made of premium polypropylene to maximize centrifugal strength and maintain clarity. Tubes and caps may be frozen at temperatures as low as 86˚C. Caps have a flat top surface for easy marking and labeling.

Features and Benefits

  • Sterile, RNase, DNase, pyrogen and cytotoxin free
  • Available racked or bagged
  • Compatible with standard rotors
  • Frosted marking surface

Legal Information

MTC is a trademark of MTC Bio


Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library