MAK335
Succinate Assay Kit
sufficient for 100 colorimetric or fluorometric tests
Synonym(s):
Succinate Quantification Kit
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12161503
NACRES:
NA.84
Recommended Products
usage
sufficient for 100 colorimetric or fluorometric tests
input
beverage(s)
food(s)
biological sample(s)
detection method
colorimetric
fluorometric
relevant disease(s)
cardiovascular diseases; immunological diseases
storage temp.
−20°C
General description
Succinate, or succinic acid, can be found in all plants and animal tissues. It is an intermediate in the citric acid cycle and plays an important role in intracellular energy generation. Succinate is widely used as a flavoring agent in the food, beverage, and pharmaceutical industries due to its low toxicity.
Application
The Succinate Assay Kit may be used for:
- Diabetes Research
- Food and Beverage Testing
- Mitochondrial Disorder Research
Features and Benefits
Efficient and Precise: Detect Succinate levels ranging from 10 to 400 μM (colorimetric) and 2 to 40 μM (fluorimetric) in a 30 minute room temperature reaction, providing both convenience and sensitivity.
Compatibility with High-Throughput Systems: Easily incorporate our kit into high-throughput handling systems, ensuring smooth and accurate processing, enhancing efficiency in your laboratory workflow.
Compatibility with High-Throughput Systems: Easily incorporate our kit into high-throughput handling systems, ensuring smooth and accurate processing, enhancing efficiency in your laboratory workflow.
Suitability
Suitable for succinate determination in food, beverage, agricultural products, and other biological samples.
Principle
The Succinate Assay Kit provides a simple, one step assay for measuring succinate. In this assay succinate is converted to pyruvate which reacts with specific reagents and dye to form a colored product. The color intensity at 570 nm or fluorescence at λex/em = 530/585 nm of the reaction product is directly proportional to succinate concentration in the sample. Linear detection range 10 μM to 400 μM for colorimetric assays and 2 to 40 μM for fluorimetric assays.
Other Notes
For additional information on our range of Biochemicals, please complete this form.
Storage Class Code
10 - Combustible liquids
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Sulagna Bhattacharya et al.
EBioMedicine, 68, 103418-103418 (2021-06-09)
We have previously described an evolutionarily selected Tibetan prolyl hydroxylase-2 (PHD2D4E;C127S) variant that degrades the hypoxia-inducible factor (HIFα) more efficiently and protects these highlanders from hypoxia-triggered elevation in haemoglobin concentration. High altitude is known to cause acute mountain sickness (AMS)
Nishith M Shrimali et al.
EBioMedicine, 73, 103672-103672 (2021-11-06)
Phospho-Akt1 (pAkt1) undergoes prolyl hydroxylation at Pro125 and Pro313 by the prolyl hydroxylase-2 (PHD2) in a reaction decarboxylating α-ketoglutarate (αKG). We investigated whether the αKG supplementation could inhibit Akt-mediated activation of platelets and monocytes, in vitro as well as in
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service