Skip to Content
Merck

HS15990B

Slide box

25 place, red

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NB.17
UNSPSC Code:
41123302
Manufacturer/tradename:
Heathrow Scientific HS15990B
Feature:
Holds 25 x slides 76x26 mm
Material:
polypropylene
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

polypropylene

feature

Holds 25 x slides 76x26 mm

manufacturer/tradename

Heathrow Scientific HS15990B

size

141x92x30 mm

color

red

Features and Benefits

  • The slide box is made of durable polypropylene.
  • The box can hold either 25 or 100 standard slides (25 x 75 mm and 1 x 3 inches).
  • Latch locks keep hinged lids securely closed.
  • Stability is maximized by stacking feet.
  • Slide identification card - capacity 100 slides
  • No liner


Still not finding the right product?

Explore all of our products under Slide box




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library