HS120652
Reagent Reservoir
capacity 25 mL, polystyrene, sterile; electron beam irradiated, pack of 100 (individually wrapped)
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
UNSPSC Code:
41121522
NACRES:
NB.01
Recommended Products
material
V-bottom bottom
polystyrene
sterility
sterile; electron beam irradiated
feature
Divided as 10 and 15 mL
packaging
pack of 100 (individually wrapped)
manufacturer/tradename
Heathrow Scientific 120652
capacity
25 mL
Looking for similar products? Visit Product Comparison Guide
General description
Reagent Reservoirs offer a fresh and modern design with enhanced features to streamline pipetting procedures. Graduation markingson the top edge and inner walls allows measurements of liquid to be easily identified and reduces waste of expensive reagents. Pour-off spouts design on all four corners enables easy spill-free pouring and reduces chance of dripping.
Features and Benefits
- Trough design with a “v” shaped bottom helps ensure that all the reagent is accessible and can be easily aspirated
- Thick wall construction and wide base adds stability and durability making the reservoir easy to handle without product deformation which results in less spillage
- Reservoirs can be conveniently stacked with a unique nesting design that enables easy separation
- Designed to stay flat on the benchtop and not warp
- Sealed in easy-to-open bags. No scissors needed to open
- Disposable
- Design Patent Pending
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service