GBL247865
Grace Bio-Labs ProPlate® tray sets
size 64 wells, with Delrin® snap clips
Synonym(s):
multi array slide system, multi slide module system
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
UNSPSC Code:
41122600
NACRES:
NB.15
material:
black
silicone (on Delrin)
square wells
silicone (on Delrin)
square wells
size:
64 wells
Recommended Products
material
black
silicone (on Delrin)
square wells
sterility
non-sterile
manufacturer/tradename
Grace Bio-Labs 247865
size
64 wells
well area
0.13 cm2
well volume
350 μL
color
none
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Grace Bio-Labs ProPlate® multi-array chamber tray sets are designed to be used with ONCYTE® nitrocellulose-coated slides. These multi-array chambers integrate microscope slide based microarray technology with high-throughput (HTS applications) microtiter plate processing. Up to 64 individual arrays can be assayed per slide.
Three or four slides fit into a tray (depending on clip choice). The resulting multi-array chamber tray set produces a modular plate with a standard microtiter plate footprint and well spacing allowing high-throughput robotic processing of up to 256 arrays.
Features:
16 or 64 well ProPlate® tray sets include the following:
Three or four slides fit into a tray (depending on clip choice). The resulting multi-array chamber tray set produces a modular plate with a standard microtiter plate footprint and well spacing allowing high-throughput robotic processing of up to 256 arrays.
Features:
- Up to 16 or 64 leak-proof wells on any slide surface without the use of adhesive
- Microtiter plate well spacing facilitates use with multichannel pipettes
- 7 x 7 mm square wells are ideal for high content arrays 5-350 uL well volumes accommodate small reagent and large wash volumes
16 or 64 well ProPlate® tray sets include the following:
- 7 x 7 mm well dimensions (x 4)
- 16 or 64 well capacity
- Stainless steel Delrin® snap clips (x 8)
- Tray & cover included (x 1)
- 10 seal strips (x 10) and 1 seal strip applicator (x 1)
Legal Information
Delrin is a registered trademark of E. I. du Pont de Nemours and Company
Oncyte is a registered trademark of Grace Bio-Labs, Inc.
ProPlate is a registered trademark of Grace Bio-Labs, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service