CLS354013
Corning® Falcon® Sample Container
capacity 110 mL, lid, sterile
Synonym(s):
Corning container
About This Item
Recommended Products
material
polypropylene
sterility
sterile
feature
lid
manufacturer/tradename
Corning 354013
capacity
110 mL
Looking for similar products? Visit Product Comparison Guide
Application
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service