CLS21502L
Pyrex® single cavity chromatography reservoir
capacity 2 L
Synonym(s):
storage reservoir
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
Pyrex
colorless
grade
for chromatography
sterility
non-sterile
packaging
pack of 1 ea
manufacturer/tradename
Corning 2150-2L
capacity
2 L
Looking for similar products? Visit Product Comparison Guide
Legal Information
PYREX is a registered trademark of Corning, Inc.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service