Skip to Content
Merck
All Photos(1)

Documents

DMS 92

95062825, human lung (bone marrow metastasis), Not specified

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

DMS 92, 95062825

biological source

human lung (bone marrow metastasis)

growth mode

Aggregates in suspension

karyotype

Modal no. 44

morphology

Not specified

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

metastasis

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human Caucasian lung small cell carcinoma, metastasis to bone marrow

Cell Line Description

DMS 92 was obtained from a bone marrow aspirate of an untreated 39 year old male at time of diagnosis. The isolated cells were identified as small cell carcinoma of the lung (SCLC). DMS 92 cells secrete adrenocorticotropin (ACTH), bombesin (BN), oxytocin-neurophysin (OT-NP) and 17 β-estradiol. Cells produce tumours in athymic nude mice.

Application

Human lung tumour model

Culture Medium

Waymouth′s MB 752/1 + 2mM Glutamine + 10% Foetal Bovine Serum (FBS) (Heat Inactivated).

Subculture Routine

Maintain cultures between 3-9x100,000 cells/ml; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service