KYSE-410
94072023, human esophagus, Epitheloid
Synonym(s):
KYSE0410 Cells, KYSE410 Cells
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41106514
Recommended Products
Product Name
KYSE-410, 94072023
biological source
human esophagus
growth mode
Adherent
karyotype
Hypotetraploid with 4% polyploidy
morphology
Epitheloid
products
Not specified
receptors
Not specified
technique(s)
cell culture | mammalian: suitable
relevant disease(s)
cancer
shipped in
dry ice
storage temp.
−196°C
Cell Line Origin
Human oesophageal squamous cell carcinoma
Cell Line Description
Established from the poorly differentiated invasive oesophageal squamous cell carcinoma resected from the cervical oesophagus of a 51-year old Japanese man prior to treatment. Reported to overexpress heparin binding growth factor (hst-1) and cyclin D. This cell line has been mycoplasma eradicated at ECACC. By short tandem repeat (STR)-PCR analysis the Y chromosome could not be detected in this cell line. It is a known phenomenon that SSC cell lines can lose their Y chromosome.
DNA Profile
STR-PCR Data: Amelogenin: X
CSF1PO: 12
D13S317: 11
D16S539: 10,12
D5S818: 13
D7S820: 12
THO1: 8
TPOX: 8,11
vWA: 16,18
CSF1PO: 12
D13S317: 11
D16S539: 10,12
D5S818: 13
D7S820: 12
THO1: 8
TPOX: 8,11
vWA: 16,18
Culture Medium
RPMI 1640 + 2mM Glutamine + 5-10% Foetal Bovine Serum (FBS)
Subculture Routine
Split confluent cultures 1:5 to 1:10 twice a week i.e. seeding at 2-4x10,000 cells/cm2 using trypsin/EDTA;5% CO2; 37°C. Doubling time ~45h
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service