BRIN-BG5
10033001, rat pancreas (islets), Epitheloid
Synonym(s):
BRIN
About This Item
product name
BRIN-BG5, 10033001
biological source
rat pancreas (islets)
growth mode
Adherent
karyotype
Modal chromosome number 66 up to passage 50
morphology
Epitheloid
products
BRIN-BG5 is a stable, glucose-responsive insulin secreting beta cell line expressing insulin, glucokinase and IAPP as detected by immunocytochemistry. BRIN-BG5 cells express the GLUT2 glucose transporter.
receptors
Not specified
technique(s)
cell culture | mammalian: suitable
shipped in
dry ice
Looking for similar products? Visit Product Comparison Guide
Cell Line Origin
Cell Line Description
Culture Medium
Subculture Routine
Other Notes
Disclaimer
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service