Skip to Content
Merck

Mero-41

NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA, 9100103, human lung, Epithelial

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41106514
Biological source:
human lung
Relevant disease(s):
cancer
Growth mode:
Adherent
Karyotype:
Number of chromosomes 68-75
Morphology:
Epithelial
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

human lung

packaging

tube of 5 μg 09100103-DNA-5UG, pkg of vial of cells 09100103-1VL

growth mode

Adherent

karyotype

Number of chromosomes 68-75

morphology

Epithelial

products

Cytokeratins 7, 8, 18 and 19. Epithelial membrane antigen (EMA)

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Application

Investigation of the properties of malignant mesothelioma cells and the development of new diagnostic methods

Biochem/physiol Actions

A human mesothelioma cell line derived from a malignant mesothelioma. Asbestos exposure
Mero-41 was derived from the pleural effusion from a male patient aged >60 with a history of asbestos exposure
STR-PCR Data: Amelogenin: X,Y
CSF1PO: 10,11
D13S317: 13
D16S539: 12
D5S818: 13
D7S820: 9
THO1: 6,8
TPOX: 8
vWA: 17

Preparation Note

Hams F10 + 15% FCS + 2mM Glutamine
Split sub-confluent cultures (70-80%) 1:4 to 1:10 i.e. seeding at 2-4x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
This cell line is part of the European Collection of Authenticated Cell Cultures (ECACC), an international repository managed by the United Kingdom Health Security Agency (UKHSA). No licensing agreement is required when either this cell line or the DNA extracted from it are used for internal research purposes only. Any other use of these products is prohibited without the express written permission of UKHSA. Inquiries regarding authorized use of this cell line or its genomic DNA may be directed to [email protected].




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library