Skip to Content
Merck
All Photos(1)

Key Documents

833KE

6072611, human embryo (carcinoma), Not specified

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

Product Name

833KE, 06072611

biological source

human embryo (carcinoma)

growth mode

Adherent

karyotype

Not specified

morphology

Not specified

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human embryonal carcinoma

Cell Line Description

A human embryonal carcinoma cell line derived from a human testicular germ cell tumour.

Culture Medium

DMEM (High Glucose) + 2mM Glutamine + 10% Foetal Bovine Serum (FBS). No sodium pyruvate.

Subculture Routine

Split sub-confluent cultures (70-80%), approximately every 3-4 days, 1:3 to 1:5 i.e. seeding at >6x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 10% CO2; 37°C. Cells need to be maintained at a high cell density.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service